Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Colour Standards, Saybolt

Saybolt colour standard +25, 500 mL
Katalog # PROOALK-SAYB02
Leverandør:   LGC Standards PROMOCHEM
Colour Standards, Saybolt
Katalog # PROOALK-SAYB02
CAS nummer:  
Colour Standards, Saybolt

Saybolt colour standard +25, 500 mL

CAS nummer:  
Price
2.800,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

This product is intended for the calibration and/or verification of equipment used for the measurement of colour of liquid samples. This product may also be used for visual based methodology.

  • Single solution
  • Store at room temperature
  • Laboratory use

Certifications: ISO 17025, ISO 17034 Certified.

Specifikationer

Specifikationer

  • Pakn:
    500 mL
  • Beskrivelse:
    Saybolt colour standard +25

Specifikationer