Computers for Laboratory Equipment
PC Dell Optiplex XE4 Tower - Processor Intel Core i5-12500 (6C - Up to 4,60 GHz); 8 GB DDR4 Memory - 512 GB SSD; VGA Port - 2X Serial Ports - DVD+/-RW; 2,5 Gb PCIe card (2nd LAN); Mouse and International English keyboard; Dell 3 years ProSupport Next Business Day On-Site warranty; 2 years Full LabService support; 12 MUI languages preloaded (EN, DE, FR, IT, NL, SE, DK, SU, NO, ES, PT, PL); Windows 11 IoT Ent. 2024 LTSC Value license operating system
Katalog # 906-0249
Leverandør: COLLECTION PREFIX HPLC
Product Details & Documents
PC equipment optimised for use with laboratory devices and software. Designed to meet laboratory conditions, ensuring necessary performance. Includes a wide range of accessories such as monitors and keyboards.
- Pre-installed with Windows LTSC 10 or 11 to reduce downtime from frequent updates and increase stability
- Compact footprint with integrated PC-monitor solutions
- Pre-installed language packs
- Keyboards with all common European language layouts
- Compatible with main CDS (Chromatography Data Systems)
Specifikationer
- Beskrivelse:PC Dell Optiplex XE4 Tower - Processor Intel Core i5-12500 (6C - Up to 4,60 GHz); 8 GB DDR4 Memory - 512 GB SSD; VGA Port - 2X Serial Ports - DVD+/-RW; 2,5 Gb PCIe card (2nd LAN); Mouse and International English keyboard; Dell 3 years ProSupport Next Business Day On-Site warranty; 2 years Full LabService support; 12 MUI languages preloaded (EN, DE, FR, IT, NL, SE, DK, SU, NO, ES, PT, PL); Windows 11 IoT Ent. 2024 LTSC Value license operating system
- Seq:08
- Cat. No.:906-0249