Computers for Laboratory Equipment
HP® printer, 8132E, colour
Katalog # 906-0257
Leverandør: COLLECTION PREFIX HPLC
Product Details & Documents
PC equipment optimised for use with laboratory devices and software. Designed to meet laboratory conditions, ensuring necessary performance. Includes a wide range of accessories such as monitors and keyboards.
- Pre-installed with Windows LTSC 10 or 11 to reduce downtime from frequent updates and increase stability
- Compact footprint with integrated PC-monitor solutions
- Pre-installed language packs
- Keyboards with all common European language layouts
- Compatible with main CDS (Chromatography Data Systems)
Specifikationer
- Beskrivelse:HP® printer, 8132E, colour
- Seq:34