Storage and Assay Microplates
Storage/reaction microplate, 96 round well, round bottom style, 32 mm height, -, 1,28 ml (max.)
Katalog # AGLS203426-100
Leverandør: AGILENT
Gå til:
Product Details & Documents
For biological assays, low storage temperatures, and filtrate collection applications.
- Compatible with all major automation platforms and benchtop devices
- No risk feasibility testing – test a sample in your application
- Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
- Molded with chemical resistant polymers, for maintained performance with prolonged exposure to aggressive solutions and solvents
- Sealable with heat and adhesive seals, or cap mats, for superior performance
- Low temperature tolerant for refrigerator or freezer storage
- Autoclavable, can be sterilized at 121 °C, 1,03 bar for 20 minutes
- Available in a large range of well volumes, from 58 to 10 ml for application scale-up or scale-down
- Complementary fit to Agilent filter microplates for centrifugation stacking
- Manufactured in a DNAse/RNase-free ISO 9001 regulated facility
Agilent supplies collection and storage assay microplates for a wide range of reaction chemistries, biological assays, low storage temperatures, and filtrate collection applications. These microplates are molded with medical-grade, chemical-resistant polymers and are available in 24, 48, 96, or 384-well configurations of variable volumes. Agilent also supplies mat seals and lids for these microplates.
Specifikationer
- Steril:-
- Beskrivelse:Storage/reaction microplate, 96 round well, round bottom style, 32 mm height
- Seq:02
- Brøndvolumen:1,28 ml (max.)
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



