Orders LinkContactUsLinkComponent
Cold trap, type KF54V
Cold trap, type KF54V
Katalog # KGWA17109
Leverandør:   KGW
Cold trap, type KF54V
Katalog # KGWA17109
Leverandør:  KGW
Cold trap, type KF54V
Cold trap, type KF54V
Price
12.150,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask
Specifikationer
  • Seq:
    01