Cold trap, type KF54V
Katalog # KGWA17109
Leverandør: KGW
Gå til:
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents.
- Easy to clean
- No stand material for holding the cold trap
- Protective casing of Dewar flask
Specifikationer
- Seq:01