Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Recombinant Human MAP2 Projection P2

Recombinant Human MAP2 Projection P2

Vial
Katalog # ANTIA270589-50
Leverandør:   Antibodies.com
Recombinant Human MAP2 Projection P2
Katalog # ANTIA270589-50
Leverandør:  Antibodies.com
CAS nummer:  
Recombinant Human MAP2 Projection P2

Vial

CAS nummer:  
Price
7.500,00 kr
Log ind for tilgængelighed
1 * 50 µl
Product Details & Documents
Specifikationer

Specifikationer

  • Pakning:
    Vial
  • Cat. No.:
    ANTIA270589-50
  • Bærerfri:
    N
  • Gen ID:
    UniprotID# P11137
  • Protein/peptide type:
    Recombinant
  • Sekvens:
    SVHESIDTMSPMHKNGDKEFQTGKESQPSPPAQEAGYSTLAQSYPSDLPEEPSSPQERMFTIDPKVYGEKRDLHSKNKDDLTLSRSLGLGGRSAIEQRSMSINLPMSCLDSIALGFNFGRGHDLSPLASDILTNTSGSMDEGDDYLPATTPALEKAPCFPVESKEEEQIEKVKATGEESTQAEISCESPFLAKDFYKNGTVMAPDLPEMLDLAGTRSRLASVSADAEVARRKSVPSETVVEDSRTGLPPVTDENHVIVKTDSQLEDLGYCVFNKYTVPLPSPVQDSENLSGESGTFYEGTDDKVRRDLATDLSLIEVKLAAAGRVKDEFSVDKEASAHISGDKSGLSKEFDQEKKANDRLDTVLEKSEEHADSKEHAKKTEEAGDEIETFGLGVTYEQALAKDLSIPTDASSEKAEKGLSSVPEIAEVEPSKKVEQGLDFAVQGQLDVKISDFGQMASGLNIDDRRATELKLEATQDMTP
  • Konjugation:
    Unconjugated
  • Protein/peptide name:
    MAP2 Projection P2
  • Tested applications:
    SDS-PAGE, ELISA
  • Species:
    Human
  • Shipping temperature:
    Shipped on blue ice at 4 °C.

Specifikationer