Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Recombinant Human MAP2 Projection P2

Recombinant Human MAP2 Projection P2

Vial
Katalog # ANTIA270589-25
Leverandør:   Antibodies.com
Recombinant Human MAP2 Projection P2
Katalog # ANTIA270589-25
Leverandør:  Antibodies.com
CAS nummer:  
Recombinant Human MAP2 Projection P2

Vial

CAS nummer:  
Price
4.480,00 kr
Log ind for tilgængelighed
1 * 25 µl
Product Details & Documents
Specifikationer

Specifikationer

  • Pakning:
    Vial
  • Cat. No.:
    ANTIA270589-25
  • Bærerfri:
    N
  • Gen ID:
    UniprotID# P11137
  • Protein/peptide type:
    Recombinant
  • Sekvens:
    SVHESIDTMSPMHKNGDKEFQTGKESQPSPPAQEAGYSTLAQSYPSDLPEEPSSPQERMFTIDPKVYGEKRDLHSKNKDDLTLSRSLGLGGRSAIEQRSMSINLPMSCLDSIALGFNFGRGHDLSPLASDILTNTSGSMDEGDDYLPATTPALEKAPCFPVESKEEEQIEKVKATGEESTQAEISCESPFLAKDFYKNGTVMAPDLPEMLDLAGTRSRLASVSADAEVARRKSVPSETVVEDSRTGLPPVTDENHVIVKTDSQLEDLGYCVFNKYTVPLPSPVQDSENLSGESGTFYEGTDDKVRRDLATDLSLIEVKLAAAGRVKDEFSVDKEASAHISGDKSGLSKEFDQEKKANDRLDTVLEKSEEHADSKEHAKKTEEAGDEIETFGLGVTYEQALAKDLSIPTDASSEKAEKGLSSVPEIAEVEPSKKVEQGLDFAVQGQLDVKISDFGQMASGLNIDDRRATELKLEATQDMTP
  • Konjugation:
    Unconjugated
  • Protein/peptide name:
    MAP2 Projection P2
  • Tested applications:
    SDS-PAGE, ELISA
  • Species:
    Human
  • Shipping temperature:
    Shipped on blue ice at 4 °C.

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.