Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant MAP2 Projection P1

Human Recombinant MAP2 Projection P1

Vial
Katalog # ANTIA270588-50
Leverandør:   Antibodies.com
Human Recombinant MAP2 Projection P1
Katalog # ANTIA270588-50
Leverandør:  Antibodies.com
CAS nummer:  
Human Recombinant MAP2 Projection P1

Vial

CAS nummer:  
Price
7.500,00 kr
Log ind for tilgængelighed
1 * 50 µl
Product Details & Documents

Partial length Human MAP2 Projection P1 suitable for SDS-PAGE, ELISA, Immunogen Grade.

Advarsel: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Specifikationer

Specifikationer

  • Pakning:
    Vial
  • Cat. No.:
    ANTIA270588-50
  • Bærerfri:
    N
  • Gen ID:
    UniprotID# P11137
  • Protein/peptide type:
    Recombinant
  • Sekvens:
    DLLTASKMEFHDQQELTPSTAEPSDQKEKESEKQSKPGEDLKHAALVSQPETTKTYPDKKDMQGTEEEKAPLALFGHTLVASLEDMKQKTEPSLVVPGIDLPKEPPTPKEQKDWFIEMPTEAKKDEWGLVAPISPGPLTPMREKDVFDDIPKWEGKQFDSPMPSPFQGGSFTLPLDVMKNEIVTETSPFAPAFLQPDDKKSLQQTSGPATAKDSFKIEEPHEAKPDKMAEAPPSEAMTLPKDAHIPVVEEHVMGKVLEEEKEAINQETVQQRDTFTPSGQEPILTEKETELKLEEKTTISDKEAVPKESKPPKPADEEIGIIQTSTEHTFSEQKDQEPTTDMLKQDSFPVSLEQAVTDSAMTSKTLEKAMTEPSALIEKSSIQELFEMRVDDKDKIEGVGAATSAELDMPFYEDKSGMSKYFETSALKEEATKSIEPGSDYYELSDTRE
  • Konjugation:
    Unconjugated
  • Protein/peptide name:
    MAP2 Projection P1
  • Molekylvægt:
    50 kDa (75 kDa by SDS-PAGE)
  • Formulering:
    Supplied in 6 M Urea.
  • Tested applications:
    SDS-PAGE, ELISA, Immunogen Grade
  • Species:
    Human
  • Shipping temperature:
    Shipped on blue ice at 4 °C.

Specifikationer