Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant MAP2 Projection P1

Human Recombinant MAP2 Projection P1

Vial
Katalog # ANTIA270588-25
Leverandør:   Antibodies.com
Human Recombinant MAP2 Projection P1
Katalog # ANTIA270588-25
Leverandør:  Antibodies.com
CAS nummer:  
Human Recombinant MAP2 Projection P1

Vial

CAS nummer:  
Price
4.480,00 kr
Log ind for tilgængelighed
1 * 25 µl
Product Details & Documents

Partial length Human MAP2 Projection P1 suitable for SDS-PAGE, ELISA, Immunogen Grade.

Advarsel: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Specifikationer

Specifikationer

  • Pakning:
    Vial
  • Cat. No.:
    ANTIA270588-25
  • Bærerfri:
    N
  • Gen ID:
    UniprotID# P11137
  • Protein/peptide type:
    Recombinant
  • Sekvens:
    DLLTASKMEFHDQQELTPSTAEPSDQKEKESEKQSKPGEDLKHAALVSQPETTKTYPDKKDMQGTEEEKAPLALFGHTLVASLEDMKQKTEPSLVVPGIDLPKEPPTPKEQKDWFIEMPTEAKKDEWGLVAPISPGPLTPMREKDVFDDIPKWEGKQFDSPMPSPFQGGSFTLPLDVMKNEIVTETSPFAPAFLQPDDKKSLQQTSGPATAKDSFKIEEPHEAKPDKMAEAPPSEAMTLPKDAHIPVVEEHVMGKVLEEEKEAINQETVQQRDTFTPSGQEPILTEKETELKLEEKTTISDKEAVPKESKPPKPADEEIGIIQTSTEHTFSEQKDQEPTTDMLKQDSFPVSLEQAVTDSAMTSKTLEKAMTEPSALIEKSSIQELFEMRVDDKDKIEGVGAATSAELDMPFYEDKSGMSKYFETSALKEEATKSIEPGSDYYELSDTRE
  • Konjugation:
    Unconjugated
  • Protein/peptide name:
    MAP2 Projection P1
  • Molekylvægt:
    50 kDa (75 kDa by SDS-PAGE)
  • Formulering:
    Supplied in 6 M Urea.
  • Tested applications:
    SDS-PAGE, ELISA, Immunogen Grade
  • Species:
    Human
  • Shipping temperature:
    Shipped on blue ice at 4 °C.

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.