Orders LinkContactUsLinkComponent
Engangsørepropper, Bilsom 303
Engangsørepropper, Bilsom 303
Earplug dispenser HL400 with 400 plugs, gul, S
Katalog # DALL1006204-AM
Leverandør:   HONEYWELL SAFETY
Engangsørepropper, Bilsom 303
Katalog # DALL1006204-AM
Leverandør:  HONEYWELL SAFETY
Engangsørepropper, Bilsom 303
Engangsørepropper, Bilsom 303
Earplug dispenser HL400 with 400 plugs, gul, S
Price
921,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Formstøbt skumpropper uden PVC, fås i to størrelser for at sikre korrekt og komfortabel pasform.
  • Polyurethane foam, without cord
  • Conical shape for easy insertion
  • Smooth outer surface for better hygiene
  • Transparent dispenser for convenience
SNR: 33 dB
EN 352-2
 : EN 352-2.
 : Available bulk packed, as individual pairs or as 400 pairs within the LS-400 dispenser.
Specifikationer
  • Farve:
    gul
  • Beskrivelse:
    Earplug dispenser HL400 with 400 plugs
  • Seq:
    01
  • Størrelse:
    S
Accessories
Produktinformation
  • Katalog #
    Farve
    Tilgængelighed
    Price
  • Ear plug dispenser for use with earplugs
    Ear plug dispenser for use with earplugs
    DALL1006203-AM
    sort
    Log ind for tilgængelighed
    1 * 1 Items - kr921.00