Magicard™ Prima 8 Clear Laminate Patch Films
Clear patch laminate for Prima 8 laminator, Laminated film, 0,6 mil
Katalog # BRDY321993
Leverandør: Brady
Gå til:
Product Details & Documents
Prima 8 Clear laminate patch film for Prima 8 laminator.
- Provide extra protection against scratches on your card
- Can be applied with Prima 8 laminator
One laminate roll.
Specifikationer
- Beskrivelse:Clear patch laminate for Prima 8 laminator
- Materiale:Laminated film
- Seq:01
- Tykkelse:0,6 mil