Orders LinkContactUsLinkComponent
Enriched Lotion™ Hand Washes
Enriched Lotion™ Hand Washes
Enriched Lotion™ hand wash for FLEX™ 1300 ml dispenser, 1300 ml
Katalog # RUBB3486616
Enriched Lotion™ Hand Washes
Katalog # RUBB3486616
Enriched Lotion™ Hand Washes
Enriched Lotion™ Hand Washes
Enriched Lotion™ hand wash for FLEX™ 1300 ml dispenser, 1300 ml
Price
130,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
High-quality Enriched Lotion™ hand wash soap leaves the skin feeling smooth and conditioned.
  • Hygienically sealed refill eliminates the potential for contamination
  • High capacity refills deliver up to 3250 hand washes for best cost in use
  • Fresh fruity fragrance appeals to all
Dermatologically tested and enriched with skin conditioners.
Specifikationer
  • Kapacitet:
    1300 ml
  • Beskrivelse:
    Enriched Lotion™ hand wash for FLEX™ 1300 ml dispenser
  • Seq:
    01
Complementary Products
Produktinformation
  • Katalog #
    Beskrivelse
    Kapacitet
    Farve
    Tilgængelighed
    Price
  • RUBBRSZ5607
    Manual spray dispenser
    400 ml
    hvid/grå
    Log ind for tilgængelighed
    1 * 1 Items - 434,00 kr