Orders LinkContactUsLinkComponent
Enriched Foam™ Hand Washes
Enriched Foam™ Hand Washes
Enriched Foam™ hand wash for FLEX™ 500 ml dispenser, 500 ml
Katalog # RUBB3486605
Enriched Foam™ Hand Washes
Katalog # RUBB3486605
Enriched Foam™ Hand Washes
Enriched Foam™ Hand Washes
Enriched Foam™ hand wash for FLEX™ 500 ml dispenser, 500 ml
Price
79,90 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Light, floral, and aromatic fragranced formula with enriched moisturisers to help prevent skin dryness.
  • Hygienically sealed refill eliminates the potential for contamination
  • Dermatologically tested and enriched with skin conditioners
  • High capacity refills deliver up to 3250 hand washes for best cost in use
  • Appeals to all, ideal for high traffic environments
Specifikationer
  • Kapacitet:
    500 ml
  • Beskrivelse:
    Enriched Foam™ hand wash for FLEX™ 500 ml dispenser
  • Seq:
    03
Complementary Products
Produktinformation
  • Katalog #
    Beskrivelse
    Kapacitet
    Farve
    Tilgængelighed
    Price
  • RUBBFG450017
    Manual foam dispenser
    800 ml
    hvid/grå
    Log ind for tilgængelighed
    1 * 1 Items - 463,00 kr