Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant FGF-16

Human Recombinant FGF-16

50 µG
Catalog # STMC78175.1
Marketsource
Human Recombinant FGF-16
Catalog # STMC78175.1
Marketsource
CAS Number:  
Human Recombinant FGF-16
Human Recombinant FGF-16

50 µG

CAS Number:  
Price
2.356,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Fibroblast growth factor 16 (FGF-16) is a heparin-binding member of the FGF family. FGFs possess broad mitogenic and cell survival activities and are expressed during embryonic development. FGFs act primarily on cells of mesodermal and neuroectodermal origin to regulate diverse physiological functions including angiogenesis, cell growth, pattern formation, embryonic development, metabolic regulation, cell migration, and tissue repair (Goldfarb; Green et al.). In vitro, FGF-16 has been shown to promote the proliferation of brown adipocytes and in rat embryos it is predominantly expressed in these cells. FGF-16 has also been shown to play a critical role in embryonic heart development and is thought to play a cardioprotective role after birth (Hotta et al.; Lu et al.; Wang et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pakn:
    50 µG
  • Cat. No.:
    STMC78175.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    FGF-16
  • Renhed:
    ≥95%
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.