Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant IGF-I, ACF

Human Recombinant IGF-I, ACF

1.000 µG
Katalog # STMC78142.2
Marketsource
Leverandør:   Stemcell Technologies
Human Recombinant IGF-I, ACF
Katalog # STMC78142.2
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Human Recombinant IGF-I, ACF
Human Recombinant IGF-I, ACF

1.000 µG

CAS nummer:  
Price
2.199,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Insulin-like growth factor 1 (IGF-I) is a polypeptide that belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin. IGF-I binds to the IGF-I receptor and is a potent activator of the PI3K/AKT pathway and also activates ERK1/2 signaling. IGF-I is required for embryonic development, and it is produced mainly in the liver in response to a hepatocyte growth hormone. In the absence of insulin, IGF-I is necessary for the maintenance of human pluripotent stem cells (Wang et al.). Together with IL-3, IGF-I stimulates differentiation and proliferation of myeloid cells and has been shown to regulate lymphopoiesis by stimulating proliferation and differentiation of T and B cells in lymphoid organs (Heemskerk et al.). This product is animal component-free.

Advarsel: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    1.000 µG
  • Cat. No.:
    STMC78142.2
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IGF-I, ACF
  • Protein synonyms:
    IBP1|IGF-IA|IGF-IB|IGF1A|Insulin-like growth factor 1|Mechano growth factor|MGF|Somatomedin C
  • Renhed:
    ≥95%
  • Species:
    Human

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.