Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant IL-13

Human Recombinant IL-13

1.000 µG
Katalog # STMC78029.3
Marketsource
Leverandør:   Stemcell Technologies
Human Recombinant IL-13
Katalog # STMC78029.3
Leverandør:  Stemcell Technologies
Marketsource
CAS nummer:  
Human Recombinant IL-13
Human Recombinant IL-13

1.000 µG

CAS nummer:  
Price
47.157,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Interleukin 13 (IL-13) is a cytokine important in type 2 immune responses and is expressed by T helper type 2 (Th2) cells and group 2 innate lymphoid cells (ILC2s) (Pulendran and Artis). IL-13 binds a receptor composed of IL-4Ra and IL-13Ra1 or IL-13Ra2 (Wynn 2003). IL-13 receptor is expressed on B cells and promotes B cell proliferation, induces class switching to IgG4 and IgE, and functions in the recruitment and activation of IgE-producing B cells (Hershey). The receptor is also expressed on basophils, eosinophils, mast cells, endothelial cells, fibroblasts, monocytes, macrophages, respiratory epithelial cells, and smooth muscle cells (Hershey). Signaling through the IL-13 receptor activates the JAK/STAT and IRS-1/IRS-2 pathways. in vivo, IL-13 has a role in resistance to extracellular helminth parasites by regulating gastrointestinal parasite expulsion, as well as in airway hyperresponsiveness, allergic inflammation, tissue remodeling, tumor cell growth, and fibrosis (Wynn 2015). Secreted IL-13 is a protein consisting of 112 amino acids with a molecular mass of 10 kDa (Hershey). Human IL-13 is not species-specific but has greater activity on human cells compared to mouse cells (Hershey).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifikationer

Specifikationer

  • Pakn:
    1.000 µG
  • Cat. No.:
    STMC78029.3
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-13
  • Renhed:
    ≥95%
  • Species:
    Human

Specifikationer