Cyclohexan-D12 99.5 atom % D for NMR spectroscopy
Glass bottle, 25 mL
Katalog # 166280251.
Leverandør: Thermo Fisher Scientific
Cyclohexan-D12 99.5 atom % D for NMR spectroscopy
Katalog # 166280251.
Leverandør: Thermo Fisher Scientific
CAS nummer: 1735-17-7
Gå til:
Product Details & Documents
Synonymer: Cyclohexane-[²H₁₂]
- CAS nummer:1735-17-7
- Formel:C₆D₁₂
- MDL nummer:MFCD00044212
- Molecular Weight:96.07 g/mol
- Kogende Pt:78 °C (760 mmHg)
- Tæthed:0,89 g/cm³
- Flash Pt:–18 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:217-077-3
- FN:1145
- ADR:3,II
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless Liquid
- Water:≤0.03 %
- D-Enrichment:≥99.4 %
Specifikationer
- Pakning:Glass bottle
- Pakn:25 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



