Cyclohexan-D12 99.5 atom % D for NMR spectroscopy
Glass bottle, 5 mL
166280051.
: Cyclohexane-[²H₁₂]
- :1735-17-7
- :C₆D₁₂
- :MFCD00044212
- :96.07 g/mol
- :78 °C (760 mmHg)
- :0,89 g/cm³
- :–18 °C
- :Stue temperatur
- :217-077-3
- :1145
- :3,II
- Appearance:Clear colorless Liquid
- Water:≤0.03 %
- D-Enrichment:≥99.4 %
- Pakning:Glass bottle
- Pakn:5 mL
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



