Inserts
Insert 1/2, E523 for sieve trays, 7 brackets, PG 8581, PG 8591, 220×450×50 mm
MIEL5649700
: MIELE
- Beskrivelse:Insert 1/2, E523 for sieve trays, 7 brackets
- Seq:25
- B×D×H:220×450×50 mm
- Til:PG 8581, PG 8591
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
#: 20864.320
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83640.320
: VWR Chemicals
Natriumhydroxid 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
#: 31770.294
: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
#: 83645.320
: VWR Chemicals