Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for LenS3™ Multi-Angle Light-Scattering Detector

Accessories for LenS3™ Multi-Angle Light-Scattering Detector

Post column filter 0,2 µm, 13 mm PES
Katalog # TOYO0040049
Leverandør:   TOSOH Bioscience
Accessories for LenS3™ Multi-Angle Light-Scattering Detector
Katalog # TOYO0040049
Leverandør:  TOSOH Bioscience
Accessories for LenS3™ Multi-Angle Light-Scattering Detector

Post column filter 0,2 µm, 13 mm PES

Price
1.130,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Post column filter 0,2 µm, 13 mm PES
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.