Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for LenS3™ Multi-Angle Light-Scattering Detector

Accessories for LenS3™ Multi-Angle Light-Scattering Detector

Post pump filter 0,1 µm, 13 mm nylon
Katalog # TOYO0040042
Leverandør:   TOSOH Bioscience
Accessories for LenS3™ Multi-Angle Light-Scattering Detector
Katalog # TOYO0040042
Leverandør:  TOSOH Bioscience
Accessories for LenS3™ Multi-Angle Light-Scattering Detector

Post pump filter 0,1 µm, 13 mm nylon

Price
97,30 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Post pump filter 0,1 µm, 13 mm nylon
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.