Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for LenS3™ Multi-Angle Light-Scattering Detector

Accessories for LenS3™ Multi-Angle Light-Scattering Detector

Post column filter 0,1 µm, 13 mm PTFE
Katalog # TOYO0040031
Leverandør:   TOSOH Bioscience
Accessories for LenS3™ Multi-Angle Light-Scattering Detector
Katalog # TOYO0040031
Leverandør:  TOSOH Bioscience
Accessories for LenS3™ Multi-Angle Light-Scattering Detector

Post column filter 0,1 µm, 13 mm PTFE

Price
97,30 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Post column filter 0,1 µm, 13 mm PTFE
  • Seq:
    01