Orders LinkContactUsLinkComponent
Accessories for SCAT Level Controls
Accessories for SCAT Level Controls
Signal lamp for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment, cable length: 3 m; Power supply: 24 V DC EU/UK/US plug. Individually extendable by SCAT signal cable via M8 connectors
Katalog # SCTV108334-T
Leverandør:   S.C.A.T.
Accessories for SCAT Level Controls
Katalog # SCTV108334-T
Leverandør:  S.C.A.T.
Accessories for SCAT Level Controls
Accessories for SCAT Level Controls
Signal lamp for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment, cable length: 3 m; Power supply: 24 V DC EU/UK/US plug. Individually extendable by SCAT signal cable via M8 connectors
Price
1.910,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Necessary and helpful accessories for use with SCAT electronic level controls.
  • Switching cables
  • Disk sensors
  • Signal cables
  • Power supply units
Specifikationer
  • Beskrivelse:
    Signal lamp for level control, visual and acoustic signal when level is reached, with magnetic holder for individual attachment, cable length: 3 m; Power supply: 24 V DC EU/UK/US plug. Individually extendable by SCAT signal cable via M8 connectors