[EN]5 COMPONENTS: 1000MG/L IN HNO3 2% 1 * 500 mL
Katalog # CMPLRM107314L5
Leverandør: CUSTOM MADE CHEMICALS LAB
[EN]5 COMPONENTS: 1000MG/L IN HNO3 2% 1 * 500 mL
Katalog # CMPLRM107314L5
Leverandør: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Formaldehyd 4% stabilised, buffered (pH 7,0 ±0,1)
Leverandør: VWR Chemicals
Immersion oil for microscopy, Sigma-Aldrich®
Leverandør: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Leverandør: Merck
Reagents for nitrate determination using Lovibond® MD photometers
Leverandør: Lovibond
Testkits, COD kuvettetest, til Spectroquant®, Supelco®
Leverandør: Merck
Afkalker, OSTEOMOLL® og OSTEOSOFT®
Leverandør: Merck
Kjeldahl tablets, Kjeltabs
Leverandør: THOMPSON & CAPPER
Testkits, ammonium test, til Spectroquant®, Supelco®
Leverandør: Merck
Reagents for Phosphate Determination using Lovibond® MD photometers
Leverandør: Lovibond
Acid red 27 ≥85-95% (dye content), Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
Acid Red 183 30% (dye content), Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
Fast Red RC Salt, Sigma-Aldrich®
Leverandør: SIGMA ALDRICH MICROSCOPY
Natriumhydroxid 30% in water, Technical Grade
Leverandør: Thermo Fisher Scientific
Nitrite VHR L, 500 ml
Leverandør: Lovibond
GLYCINE (USP, BP, PH. EUR.) PURE, PHARMA 1 * 1.000 g
Leverandør: PanReac AppliChem