Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Millenium Grip Antislip Overshoes

Millenium Grip Antislip Overshoes

46/48, sort
Catalog # MILEMLOGN-XL
Supplier:   MILLE S.A.S
Millenium Grip Antislip Overshoes
Catalog # MILEMLOGN-XL
Supplier:  MILLE S.A.S
Millenium Grip Antislip Overshoes

46/48, sort

Price
250,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Millenium grip waterproof and anti-slip overshoes for wet and oily floor, ultra light and non-magnetic.

  • TPU waterproof overshoe
  • Easy on - easy off
  • Without toe-cap
  • Without penetration resistant midsole
  • Reusable, can by worn by more than 1 person => easy to wash
  • Exclusive design to avoid clogging with dirt
  • High performance grip, perfect for slippery, wet and oily floors
  • TPU outsole
  • Slip resistance: SRC
  • FO: Fuel oil resistant sole
  • Amagnetic (without metal)
  • Outsole abrasion and hydrocarbon resistant
  • Elasticity
  • Sole: Waterproof and washable TPU

Industry: Logistics, services, administrations, hygiene - food industry, catering.

Net weight per pair: 590 g.

Certifications: FO SRC, EN ISO 20344

Specifications
  • Skostørrelse:
    46/48
  • Farve:
    sort
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.