Millenium Grip Antislip Overshoes
34/37, sort
Katalog # MILEMLOGN-S
Leverandør: MILLE S.A.S
Gå til:
Product Details & Documents
Millenium grip waterproof and anti-slip overshoes for wet and oily floor, ultra light and non-magnetic.
- TPU waterproof overshoe
- Easy on - easy off
- Without toe-cap
- Without penetration resistant midsole
- Reusable, can by worn by more than 1 person => easy to wash
- Exclusive design to avoid clogging with dirt
- High performance grip, perfect for slippery, wet and oily floors
- TPU outsole
- Slip resistance: SRC
- FO: Fuel oil resistant sole
- Amagnetic (without metal)
- Outsole abrasion and hydrocarbon resistant
- Elasticity
- Sole: Waterproof and washable TPU
Industry: Logistics, services, administrations, hygiene - food industry, catering.
Net weight per pair: 590 g.
: FO SRC, EN ISO 20344
Specifikationer
- Skostørrelse:34/37
- Farve:sort