Accessories for epMotion®
TipHolder 73 reusable and autoclavable, epBlue 40.7 or higher required
Katalog # EPPE5075751.879
Leverandør: EPPENDORF
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:TipHolder 73 reusable and autoclavable, epBlue 40.7 or higher required
- Seq:40