Orders LinkContactUsLinkComponent
Rotational viscometers, ROTAVISC
Rotational viscometer, Me-vi, 100 - 40000000 mPas
  531-1488
 :   IKA
Rotational viscometers, ROTAVISC
  531-1488
 :  IKA
Rotational viscometers, ROTAVISC
Rotational viscometers, ROTAVISC
Rotational viscometer, Me-vi, 100 - 40000000 mPas
28.800,00 kr
1 * 1 Items
The ROTAVISC series determines the viscosity of liquids in all areas of application from the laboratory to quality control.
  • Easy to operate, highest measurement accuracy and stepless speed
  • TFT display for an intuitive menu guidance
  • Time setting range from 0,017 to 6000 min
  • Overload protection and right side rotation
  • PT100 connection for external temperature sensor
A digital spirit level also supports the correct set-up of the start-up condition and a simple ramp function simplifies repetitive tasks.
 : IP 40 protection class according to DIN EN 60529.
 : Includes a spindle set, a protective bracket, temperature sensor, quick connector, hook connector and stand, Ø 16 mm support rod with fastening.
  • Model:
    Me-vi
  • Område:
    100 - 40000000 mPas
  • Beskrivelse:
    Rotational viscometer
  • Seq:
    04
  • Cat. No.:
    531-1488
  • Driftstemperatur:
    5…40 °C
  • B×D×H:
    351×372×629 mm
  • Vægt:
    7,1 kg
  • Elektriske specifikationer:
    100 – 240 V
  • Interface:
    USB and RS232 interface
  • Display:
    TFT, 4,3"
Accessories
  •  
    Beskrivelse
  • IKAA25000352
    Adapter, VOLS-1, to measure the viscosity of especially valuable and expensive samples
    1 * 1 Each - 11.000,00 kr
  •  
    Beskrivelse
  • IKAA25000466
    Standard silicone oil, CAL-O-12500
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
  • IKAA25000387
    Adapter, DINS-1, to comply with DIN 53019 standard
    1 * 1 Each - 19.350,00 kr
  •  
    Beskrivelse
  • IKAA25000399
    Standard silicone oil, CAL-O-50
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
  • IKAA25000398
    Standard silicone oil, CAL-O-10
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
  • IKAA25000434
    Standard silicone oil, CAL-O-100
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
  • IKAA25006851
    VOL-SP-11, spindle for extended viscosity range, suitable for sample volumes of 11 ml
    1 * 1 Items - 3.130,00 kr
  •  
    Beskrivelse
  • IKAA25000436
    Standard silicone oil, CAL-O-1000
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
  • IKAA25000468
    Standard silicone oil, CAL-O-60000
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
    Model
    Temp. stabilitet
  • IKAA25004524
    HRC 2 temperature control
    HRC 2 control
    0,05±K
    1 * 1 Items - 50.850,00 kr
  •  
    Beskrivelse
  • IKAA20113825
    High-temperate system ROTATEMP, for ROTAVISC up to 300 °C, EU plug
    1 * 1 Items - 30.940,00 kr
  •  
    Beskrivelse
    Til
  • IKAA4569000
    Hose, silicone, 2×1,5 m, Ø 8 mm
    1 * 1 Items - 538,00 kr
  •  
    Beskrivelse
  • IKAA20116428
    Stand for ROTAVISC
    1 * 1 Items - 7.830,00 kr
  •  
    Beskrivelse
  • IKAA25000377
    Spiral adapter, SAS-1, to measure the viscosity of very sticky and other viscous samples
    1 * 1 Each - 10.210,00 kr
  •  
    Beskrivelse
  • IKAA25006852
    VOL-SP-13.5, spindle for extended viscosity range, suitable for sample volumes of 13,5 ml
    1 * 1 Items - 3.130,00 kr
  •  
    Beskrivelse
  • IKAA25000390
    Adapter, ELVAS-1, to measure very low viscosities
    1 * 1 Each - 12.810,00 kr
  •  
    Beskrivelse
  • IKAA25000469
    Standard silicone oil, CAL-O-100000
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
    Til
  • IKAA20019397
    Labworldsoft® 6 Starter
    Small configurations with up to 3 devices can be controlled at the same time
    1 * 1 Items - 45.630,00 kr
  •  
    Beskrivelse
  • IKAA25000437
    Standard silicone oil, CAL-O-5000
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
    Til
  • IKAA20017366
    Labworldsoft® 6 Pro, with access to the entire device library
    All configurations with any number of devices can be controlled at the same time
    1 * 1 Items - 104.110,00 kr
  •  
    Beskrivelse
  • IKAA25000407
    Vane spindles (set of 4), VAN-1, allow the viscosity measurement of suspensions
    1 * 1 Each - 9.710,00 kr
  •  
    Beskrivelse
  • IKAA20114188
    VOLS-1 flex adapter set for low volumes
    1 * 1 Items - 5.600,00 kr
  •  
    Beskrivelse
    Til
  • IKAA20101872
    Labworldsoft® 6 Visc
    Viscosity measurements for thermostats and pH meters of all manufacturers
    1 * 1 Items - 10.150,00 kr
  •  
    Beskrivelse
  • IKAA20118239
    VOL-SP-2.1 Cylinder spindle with hook suspension, 95,8 mm
    1 * 1 Items - 3.130,00 kr
  •  
    Beskrivelse
  • IKAA20128647
    HELISTAND set includes T-spindles for non-flowing samples, EU plug
    1 * 1 Items - 13.890,00 kr
  •  
    Beskrivelse
  • IKAA25000467
    Standard silicone oil, CAL-O-30000
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
    Temperaturområde
    Kapacitet
  • IKAA20102007
    CBC VISC lite temperature control, EU plug
    −25...+125 °C
    5,5 - 8,5 L
    1 * 1 Items - 31.010,00 kr
  •  
    Beskrivelse
  • IKAA25000435
    Standard silicone oil, CAL-O-500
    1 * 500 mL - 1.270,00 kr
  •  
    Beskrivelse
  • IKAA25006850
    VOL-SP-10.4, spindle for extended viscosity range, suitable for sample volumes of 10,4 ml
    1 * 1 Items - 3.130,00 kr
  •  
    Beskrivelse
  • IKAA25006849
    VOL-SP-7.1, spindle for extended viscosity range, suitable for sample volumes of 7,1 ml
    1 * 1 Items - 3.050,00 kr
  •  
    Beskrivelse
  • IKAA25000397
    Standard silicone oil, CAL-O-5
    1 * 500 mL - 1.270,00 kr