Rotational viscometers, ROTAVISC
Rotational viscometer, Me-vi, 100 - 40000000 mPas
531-1488
: IKA
The ROTAVISC series determines the viscosity of liquids in all areas of application from the laboratory to quality control.
- Easy to operate, highest measurement accuracy and stepless speed
- TFT display for an intuitive menu guidance
- Time setting range from 0,017 to 6000 min
- Overload protection and right side rotation
- PT100 connection for external temperature sensor
A digital spirit level also supports the correct set-up of the start-up condition and a simple ramp function simplifies repetitive tasks.
: IP 40 protection class according to DIN EN 60529.
: Includes a spindle set, a protective bracket, temperature sensor, quick connector, hook connector and stand, Ø 16 mm support rod with fastening.
- Model:Me-vi
- Område:100 - 40000000 mPas
- Beskrivelse:Rotational viscometer
- Seq:04
- Cat. No.:531-1488
- Driftstemperatur:5…40 °C
- B×D×H:351×372×629 mm
- Vægt:7,1 kg
- Elektriske specifikationer:100 – 240 V
- Interface:USB and RS232 interface
- Display:TFT, 4,3"
Accessories
- Beskrivelse
- IKAA25000352Adapter, VOLS-1, to measure the viscosity of especially valuable and expensive samples1 * 1 Each - 11.000,00 kr
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- Beskrivelse
- IKAA25006851VOL-SP-11, spindle for extended viscosity range, suitable for sample volumes of 11 ml1 * 1 Items - 3.130,00 kr
- Beskrivelse
- Beskrivelse
- BeskrivelseModelTemp. stabilitet
- Beskrivelse
- IKAA20113825High-temperate system ROTATEMP, for ROTAVISC up to 300 °C, EU plug1 * 1 Items - 30.940,00 kr
- BeskrivelseTil
- Beskrivelse
- Beskrivelse
- IKAA25000377Spiral adapter, SAS-1, to measure the viscosity of very sticky and other viscous samples1 * 1 Each - 10.210,00 kr
- Beskrivelse
- IKAA25006852VOL-SP-13.5, spindle for extended viscosity range, suitable for sample volumes of 13,5 ml1 * 1 Items - 3.130,00 kr
- Beskrivelse
- Beskrivelse
- BeskrivelseTil
- IKAA20019397Labworldsoft® 6 StarterSmall configurations with up to 3 devices can be controlled at the same time1 * 1 Items - 45.630,00 kr
- Beskrivelse
- BeskrivelseTil
- IKAA20017366Labworldsoft® 6 Pro, with access to the entire device libraryAll configurations with any number of devices can be controlled at the same time1 * 1 Items - 104.110,00 kr
- Beskrivelse
- IKAA25000407Vane spindles (set of 4), VAN-1, allow the viscosity measurement of suspensions1 * 1 Each - 9.710,00 kr
- Beskrivelse
- BeskrivelseTil
- IKAA20101872Labworldsoft® 6 ViscViscosity measurements for thermostats and pH meters of all manufacturers1 * 1 Items - 10.150,00 kr
- Beskrivelse
- Beskrivelse
- IKAA20128647HELISTAND set includes T-spindles for non-flowing samples, EU plug1 * 1 Items - 13.890,00 kr
- Beskrivelse
- BeskrivelseTemperaturområdeKapacitet
- IKAA20102007CBC VISC lite temperature control, EU plug−25...+125 °C5,5 - 8,5 L1 * 1 Items - 31.010,00 kr
- Beskrivelse
- Beskrivelse
- IKAA25006850VOL-SP-10.4, spindle for extended viscosity range, suitable for sample volumes of 10,4 ml1 * 1 Items - 3.130,00 kr
- Beskrivelse
- IKAA25006849VOL-SP-7.1, spindle for extended viscosity range, suitable for sample volumes of 7,1 ml1 * 1 Items - 3.050,00 kr
- Beskrivelse