Rotational viscometers, ROTAVISC
Rotational viscometer, Hi-vi I, 200 - 80000000 mPas
Katalog # 531-1489
Leverandør: IKA
Gå til:
Product Details & Documents
The ROTAVISC series determines the viscosity of liquids in all areas of application from the laboratory to quality control.
- Easy to operate, highest measurement accuracy and stepless speed
- TFT display for an intuitive menu guidance
- Time setting range from 0,017 to 6000 min
- Overload protection and right side rotation
- PT100 connection for external temperature sensor
A digital spirit level also supports the correct set-up of the start-up condition and a simple ramp function simplifies repetitive tasks.
Certifications: IP 40 protection class according to DIN EN 60529.
Leveringsinformation: Includes a spindle set, a protective bracket, temperature sensor, quick connector, hook connector and stand, Ø 16 mm support rod with fastening.
Specifikationer
- Model:Hi-vi I
- Område:200 - 80000000 mPas
- Beskrivelse:Rotational viscometer
- Seq:06
- Cat. No.:531-1489
- Driftstemperatur:5…40 °C
- B×D×H:351×372×629 mm
- Vægt:7,1 kg
- Elektriske specifikationer:100 – 240 V
- Interface:USB and RS232 interface
- Display:TFT, 4,3"
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Adapter, VOLS-1, to measure the viscosity of especially valuable and expensive samplesAdapter, VOLS-1, to measure the viscosity of especially valuable and expensive samplesIKAA25000352Log ind for tilgængelighed1 * 1 Each - 11.000,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-12500
Produktinformation
- Katalog #TilgængelighedPrice
- Adapter, DINS-1, to comply with DIN 53019 standardAdapter, DINS-1, to comply with DIN 53019 standardIKAA25000387Log ind for tilgængelighed1 * 1 Each - 19.350,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-50
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-10
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-100
Produktinformation
- Katalog #TilgængelighedPrice
- VOL-SP-11, spindle for extended viscosity range, suitable for sample volumes of 11 mlVOL-SP-11, spindle for extended viscosity range, suitable for sample volumes of 11 mlIKAA25006851Log ind for tilgængelighed1 * 1 Items - 3.130,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-1000
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-60000
Produktinformation
- Katalog #ModelTemp. stabilitetTilgængelighedPrice
- HRC 2 temperature controlHRC 2 temperature controlIKAA25004524HRC 2 control0,05±KLog ind for tilgængelighed1 * 1 Items - 50.850,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- High-temperate system ROTATEMP, for ROTAVISC up to 300 °C, EU plugHigh-temperate system ROTATEMP, for ROTAVISC up to 300 °C, EU plugIKAA20113825Log ind for tilgængelighed1 * 1 Items - 30.940,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Hose, silicone, 2×1,5 m, Ø 8 mm
Produktinformation
- Katalog #TilgængelighedPrice
- Stand for ROTAVISC
Produktinformation
- Katalog #TilgængelighedPrice
- Spiral adapter, SAS-1, to measure the viscosity of very sticky and other viscous samplesSpiral adapter, SAS-1, to measure the viscosity of very sticky and other viscous samplesIKAA25000377Log ind for tilgængelighed1 * 1 Each - 10.210,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- VOL-SP-13.5, spindle for extended viscosity range, suitable for sample volumes of 13,5 mlVOL-SP-13.5, spindle for extended viscosity range, suitable for sample volumes of 13,5 mlIKAA25006852Log ind for tilgængelighed1 * 1 Items - 3.130,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Adapter, ELVAS-1, to measure very low viscositiesAdapter, ELVAS-1, to measure very low viscositiesIKAA25000390Log ind for tilgængelighed1 * 1 Each - 12.810,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-100000Standard silicone oil, CAL-O-100000IKAA25000469Log ind for tilgængelighed1 * 500 mL - 1.270,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Labworldsoft® 6 StarterLabworldsoft® 6 StarterIKAA20019397Small configurations with up to 3 devices can be controlled at the same timeLog ind for tilgængelighed1 * 1 Items - 45.630,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-5000
Produktinformation
- Katalog #TilTilgængelighedPrice
- Labworldsoft® 6 Pro, with access to the entire device libraryLabworldsoft® 6 Pro, with access to the entire device libraryIKAA20017366All configurations with any number of devices can be controlled at the same timeLog ind for tilgængelighed1 * 1 Items - 104.110,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Vane spindles (set of 4), VAN-1, allow the viscosity measurement of suspensionsVane spindles (set of 4), VAN-1, allow the viscosity measurement of suspensionsIKAA25000407Log ind for tilgængelighed1 * 1 Each - 9.710,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- VOLS-1 flex adapter set for low volumesVOLS-1 flex adapter set for low volumesIKAA20114188Log ind for tilgængelighed1 * 1 Items - 5.600,00 kr
Produktinformation
- Katalog #TilTilgængelighedPrice
- Labworldsoft® 6 ViscLabworldsoft® 6 ViscIKAA20101872Viscosity measurements for thermostats and pH meters of all manufacturersLog ind for tilgængelighed1 * 1 Items - 10.150,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- VOL-SP-2.1 Cylinder spindle with hook suspension, 95,8 mmVOL-SP-2.1 Cylinder spindle with hook suspension, 95,8 mmIKAA20118239Log ind for tilgængelighed1 * 1 Items - 3.130,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- HELISTAND set includes T-spindles for non-flowing samples, EU plugHELISTAND set includes T-spindles for non-flowing samples, EU plugIKAA20128647Log ind for tilgængelighed1 * 1 Items - 13.890,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-30000
Produktinformation
- Katalog #TemperaturområdeKapacitetTilgængelighedPrice
- CBC VISC lite temperature control, EU plugCBC VISC lite temperature control, EU plugIKAA20102007−25...+125 °C5,5 - 8,5 LLog ind for tilgængelighed1 * 1 Items - 31.010,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-500
Produktinformation
- Katalog #TilgængelighedPrice
- VOL-SP-10.4, spindle for extended viscosity range, suitable for sample volumes of 10,4 mlVOL-SP-10.4, spindle for extended viscosity range, suitable for sample volumes of 10,4 mlIKAA25006850Log ind for tilgængelighed1 * 1 Items - 3.130,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- VOL-SP-7.1, spindle for extended viscosity range, suitable for sample volumes of 7,1 mlVOL-SP-7.1, spindle for extended viscosity range, suitable for sample volumes of 7,1 mlIKAA25006849Log ind for tilgængelighed1 * 1 Items - 3.050,00 kr
Produktinformation
- Katalog #TilgængelighedPrice
- Standard silicone oil, CAL-O-5