Orders LinkContactUsLinkComponent
Microplates, 96-well
96-well microplate, F-bottom (standard) with barcode39, MICROLON® 600, 25 - 340 µl, 382 µl
Katalog # GREI655061_NOV
   Lav fremstillingspåvirkning
Leverandør:   Greiner Bio-One
Microplates, 96-well
Katalog # GREI655061_NOV
Leverandør:  Greiner Bio-One
   Lav fremstillingspåvirkning
Microplates, 96-well
Microplates, 96-well
96-well microplate, F-bottom (standard) with barcode39, MICROLON® 600, 25 - 340 µl, 382 µl
Price
460,00 kr
Log ind for tilgængelighed
1 * 40 Items
Product Details & Documents
Crystal clear PS, without lid. Ideal for ELISA methodologies, these plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600) or medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200).
  • Available with a choice of well profile: U-bottom, V-bottom, F-bottom (standard), F-bottom (chimney well), or half area
  • Free of DNase, RNase and human DNA, and are non-pyrogenic
Specifikationer
  • Beskrivelse:
    96-well microplate, F-bottom (standard) with barcode39, MICROLON® 600
  • Seq:
    06
  • Arbejdsvolumen:
    25 - 340 µl
  • Total volumen:
    382 µl
  • Cat. No.:
    GREI655061_NOV
  • Miljøcertificering:
    ISO 14001 (241002); ISO 50001 (241002)
  • Environmentally Preferable:
    Lav fremstillingspåvirkning