0,00 kr
Ordreindtastning
/
Products
Services
Digital Solutions
Applications
Support
About
Denmark
Orders Link
ContactUsLinkComponent
Mikroskop mål
Fluorite Objectives for Revolve Microscope
Fluorite Objectives for Revolve Microscope
Plan fluorite, 0,13 NA aperture, 4×, 17 mm
ECHLCMAG0042
:
DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
ECHLCMAG0042
:
DISCOVER ECHO
Fluorite Objectives for Revolve Microscope
Plan fluorite, 0,13 NA aperture, 4×, 17 mm
4.170,00 kr
1 * 1 Items
-
+
ECHO Meet Revolve
Forstørrelse:
4×
Beskrivelse:
Plan fluorite, 0,13 NA aperture
Seq:
27
Arbejdsafstand:
17 mm
Accessories for Cellcyte Gen 2 Live Cell Imaging and Analysis
#: 630-3560
: DISCOVER ECHO