Blow lamp, X 2000
Soudogaz® Blow lamp X 2000
Katalog # CAMP2201877
Leverandør: CAMPING GAZ
Gå til:
Product Details & Documents
Multi-purpose blowtorch for soft and hard soldering and many other applications.
- Handle made of polypropylene for high impact resistance
- One-hand operation of the adjustment knob
- Gas consumption: 120 g/h
- Flame temperatures up to 1750 °C
Butane cartridge available separately.
Run time: >1 h 40 min
Weight: 380 g
Specifikationer
- Beskrivelse:Soudogaz® Blow lamp X 2000
- Seq:03
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- C206, kapacitet 190 g, til bunsenbrænder 206 og X 2000 blæselamperC206, kapacitet 190 g, til bunsenbrænder 206 og X 2000 blæselamper451-2001Log ind for tilgængelighed1 * 1 Items - 38,20 kr