Multi-Layer Foil Gas Sampling Bags
Gas sampling bags, 40 L, 610×622 mm
Katalog # REST22968
Leverandør: Restek
Gå til:
Product Details & Documents
Sampling bags are cost-effective air sampling device for high level VOC and permanent gases. It features a PP combo valve with hose connection to fit 4,76 mm ID tubing and syringe port with replaceable septum. A single eyelet provides handling convenience.
- Good stability for low molecular weight compounds, such as methane, CO, CO₂ and permanent gases
- Chemically inert with light and moisture protection
- Not recommended for low ppm VOC due to background levels
- Protective five layer barrier minimises gas permeability
- Continuous sampling temperature up to 88 °C
Documents
REST22950_TDS_EN
Datablad
Specifikationer
- Kapacitet:40 L
- Beskrivelse:Gas sampling bags
- Seq:05
- B×L:610×622 mm
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



