Nylon Wire and Cable Ties
Nylon cable ties, width 2,50 mm, ufarvet, 13,5 cm
Katalog # BRDY196307
Leverandør: Brady
Gå til:
Product Details & Documents
Specifikationer
- Farve:ufarvet
- Beskrivelse:Nylon cable ties, width 2,50 mm
- Længde:13,5 cm
- Seq:01