Pressure Transducers
Pressure transducer, ¹/₂×¹/₂" TC, polysulfone, sterile, calibrated, −10 - +75 psi
Katalog # SPECACPM-05TC-C1S
Leverandør: Spectrum Laboratories
Gå til:
Product Details & Documents
Specifikationer
- Område:−10 - +75 psi
- Beskrivelse:Pressure transducer, ¹/₂×¹/₂" TC, polysulfone, sterile, calibrated
- Seq:01
- Cat. No.:SPECACPM-05TC-C1S
- Nøjagtighed:±2% from 0 - 6 psi, ±3% from 6 - 30 psi