Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pipe Markers, Water

Pipe Markers, Water

Softend water, Polyester, 150×12 mm
  BRDYN006135
 :   Brady
Pipe Markers, Water
  BRDYN006135
 :  Brady
Pipe Markers, Water
Pipe Markers, Water

Softend water, Polyester, 150×12 mm

 
97,10 kr
   
1 * 1 Items
 

Individual markers on card with precut arrows, without pictogram.

  • Colour: White on green
  • Permanent acrylic
  • Pipe diameter: 20 to 49 mm
 
  • Beskrivelse:
    Softend water
  • Dimensioner:
    150×12 mm
  • Materiale:
    Polyester
  • Seq:
    3