Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Pipe Markers, Water

De-mineralised water, Polyester, 150×12 mm
Katalog # BRDYN005873
Leverandør:   Brady
Pipe Markers, Water
Katalog # BRDYN005873
Leverandør:  Brady
Pipe Markers, Water
Pipe Markers, Water

De-mineralised water, Polyester, 150×12 mm

Price
97,10 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Individual markers on card with precut arrows, without pictogram.

  • Colour: White on green
  • Permanent acrylic
  • Pipe diameter: 20 to 49 mm
Specifikationer
  • Beskrivelse:
    De-mineralised water
  • Dimensioner:
    150×12 mm
  • Materiale:
    Polyester
  • Seq:
    2