Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Pipe Markers, Water

Pipe Markers, Water

De-mineralised water, Polyester, 150×12 mm
  BRDYN005832
 :   Brady
Pipe Markers, Water
  BRDYN005832
 :  Brady
Pipe Markers, Water
Pipe Markers, Water

De-mineralised water, Polyester, 150×12 mm

 
97,10 kr
   
1 * 1 Items
 

Individual markers on card with precut arrows, without pictogram.

  • Colour: White on green
  • Permanent acrylic
  • Pipe diameter: 20 to 49 mm
 
  • Beskrivelse:
    De-mineralised water
  • Dimensioner:
    150×12 mm
  • Materiale:
    Polyester
  • Seq:
    1
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.