Diffusion tubes, with direct indication, for TWA measurements
Nitrogen dioxide 10/a-D, 10 - 200 ppm / 1 hr
Katalog # DRAE8101111
Leverandør: Dräger
Diffusion tubes, with direct indication, for TWA measurements
Katalog # DRAE8101111
Leverandør: Dräger
Gå til:
Product Details & Documents
Long-term measurements with diffusion tubes provide integrated measurements that represent the average concentration during the sampling period.
- Measurements performed between one and eight H
- Tubes can be used as personal, or area monitors
- Determine the weight average concentration
Specifikationer
- Område:10 - 200 ppm / 1 hr
- Beskrivelse:Nitrogen dioxide 10/a-D
- Seq:02
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Holders