Orders LinkContactUsLinkComponent
Diffusion tubes, with direct indication, for TWA measurements
Diffusion tubes, with direct indication, for TWA measurements
Nitrogen dioxide 10/a-D, 10 - 200 ppm / 1 hr
Katalog # DRAE8101111
Leverandør:   Dräger
Diffusion tubes, with direct indication, for TWA measurements
Katalog # DRAE8101111
Leverandør:  Dräger
Diffusion tubes, with direct indication, for TWA measurements
Diffusion tubes, with direct indication, for TWA measurements
Nitrogen dioxide 10/a-D, 10 - 200 ppm / 1 hr
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Long-term measurements with diffusion tubes provide integrated measurements that represent the average concentration during the sampling period.
  • Measurements performed between one and eight H
  • Tubes can be used as personal, or area monitors
  • Determine the weight average concentration
Specifikationer
  • Område:
    10 - 200 ppm / 1 hr
  • Beskrivelse:
    Nitrogen dioxide 10/a-D
  • Seq:
    02
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Holders
    Holders
    DRAE6733014
    Log ind for tilgængelighed
    1 * 3 Items - 552,00 kr