Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
4'-Demethylepipodophyllotoxin

4'-Demethylepipodophyllotoxin

Bottle, 1 g
Katalog # APOSOR3101T-1G
Leverandør:   Apollo Scientific
4'-Demethylepipodophyllotoxin
Katalog # APOSOR3101T-1G
Leverandør:  Apollo Scientific
CAS nummer:  6559-91-7
4'-Demethylepipodophyllotoxin

Bottle, 1 g

CAS nummer:  6559-91-7
Price
220,00 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

A potent inhibitor of microtubule assembly.

Synonymer:  (5R,5aR,8aR,9S)-9-Hydroxy-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
  • CAS nummer:6559-91-7
  • Formel:C₂₁H₂₀O₈
  • MDL nummer:MFCD00189421
  • Molecular Weight:400.38 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle
  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.