Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

FMOC-Asp(biotinyl-PEG)-OH

Plastic bottle, 1 g
Katalog # AATB5024
Irriterende
Leverandør:   AAT Bioquest
FMOC-Asp(biotinyl-PEG)-OH
Katalog # AATB5024
Leverandør:  AAT Bioquest
CAS nummer:  
FMOC-Asp(biotinyl-PEG)-OH
FMOC-Asp(biotinyl-PEG)-OH

Plastic bottle, 1 g

CAS nummer:  
Price
kr11,260.00
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents

This PEG biotin-labeled amino acid has excellent solubility in DMF and other solvents used in SPPS.

  • Specific reactivity
  • Stable conjugation
  • High solubility
  • ADR:5,II
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.