Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
FMOC-Asp(biotinyl-PEG)-OH

FMOC-Asp(biotinyl-PEG)-OH

Plastic bottle, 100 mg
Katalog # AATB5023
Irriterende
Leverandør:   AAT Bioquest
FMOC-Asp(biotinyl-PEG)-OH
Katalog # AATB5023
Leverandør:  AAT Bioquest
CAS nummer:  
FMOC-Asp(biotinyl-PEG)-OH
FMOC-Asp(biotinyl-PEG)-OH

Plastic bottle, 100 mg

CAS nummer:  
Price
2.350,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents

This PEG biotin-labeled amino acid has excellent solubility in DMF and other solvents used in SPPS.

  • Specific reactivity
  • Stable conjugation
  • High solubility
  • ADR:5,II
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    100 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.