Orders LinkContactUsLinkComponent
Covipyte™ ED450
1.000 Tests
  AATB13538
Covipyte™ ED450
  AATB13538
 :  
Covipyte™ ED450
Covipyte™ ED450
1.000 Tests
 :  
10.730,00 kr
1 * 1 Each
Covidyte™ ED450 is a peptide substrate containing 12 amino acid sequence (VNSTLQSGLRKM) that can be cleaved by coronavirus proteases. The dark-FRET peptide contains Dabcyl (quencher) and Edans (donor) on the C-and N-terminals respectively where the fluorescence of Edans is effectively quenched by Dabcyl when the peptide is intact. When the peptide is hydrolyzed by coronavirus proteases, the Edans fragment generates significantly enhanced fluorescence since its fluorescence is no longer quenched by Dabcyl. The activity of coronavirus proteases can be effectively monitored by the fluorescence intensity of Edans. Covidyte™ ED450 is a convenient tool for screening inhibitors of coronavirus proteases.
  • MW 2034.36
 : For research use only.
  • Pakn:
    1.000 Tests