Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Covipyte™ ED450

Covipyte™ ED450

100 Tests
Katalog # AATB13537
Leverandør:   AAT Bioquest
Covipyte™ ED450
Katalog # AATB13537
Leverandør:  AAT Bioquest
CAS nummer:  
Covipyte™ ED450
Covipyte™ ED450

100 Tests

CAS nummer:  
Price
2.240,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Covidyte™ ED450 is a peptide substrate containing 12 amino acid sequence (VNSTLQSGLRKM) that can be cleaved by coronavirus proteases. The dark-FRET peptide contains Dabcyl (quencher) and Edans (donor) on the C-and N-terminals respectively where the fluorescence of Edans is effectively quenched by Dabcyl when the peptide is intact. When the peptide is hydrolyzed by coronavirus proteases, the Edans fragment generates significantly enhanced fluorescence since its fluorescence is no longer quenched by Dabcyl. The activity of coronavirus proteases can be effectively monitored by the fluorescence intensity of Edans. Covidyte™ ED450 is a convenient tool for screening inhibitors of coronavirus proteases.

  • MW 2034.36

Advarsel: For research use only.

Specifikationer

Specifikationer

  • Pakn:
    100 Tests

Specifikationer