Orders LinkContactUsLinkComponent
ReadiUse™ Viral RNA Lysis Buffer
ReadiUse™ Viral RNA lysis buffer, 50 mL
  AATB60015
ReadiUse™ Viral RNA Lysis Buffer
  AATB60015
 :  
ReadiUse™ Viral RNA Lysis Buffer
ReadiUse™ Viral RNA Lysis Buffer
ReadiUse™ Viral RNA lysis buffer, 50 mL
 :  
1.680,00 kr
1 * 50 mL
Current demands for RT-PCR testing of SARS-CoV-2 has led to a worldwide shortage of RNA extraction kits.
  • Useful as an RNA extraction kit component
  • Ready to use
  • In-house quality control
 : For research use only.
  • Pakn:
    50 mL
  • Beskrivelse:
    ReadiUse™ Viral RNA lysis buffer