ReadiUse™ Viral RNA Lysis Buffer
ReadiUse™ Viral RNA lysis buffer, 50 mL
AATB60015
Current demands for RT-PCR testing of SARS-CoV-2 has led to a worldwide shortage of RNA extraction kits.
- Useful as an RNA extraction kit component
- Ready to use
- In-house quality control
: For research use only.
- Pakn:50 mL
- Beskrivelse:ReadiUse™ Viral RNA lysis buffer