Q4ever™ Green
Q4ever™ Green, 1250X DMSO solution, 2 mL
AATB17609
A real-time polymerase chain reaction (real-time PCR), also known as quantitative polymerase chain reaction (qPCR), is a popular laboratory technique of molecular biology based on the polymerase chain reaction (PCR).
- No probe required
- Enhanced sensitivity
- Low enzymatic inhibition
- Binds to any dsDNA
: For research use only. Store at −20 °C, protected from light. Product is stable for at least 12 months from the date of receipt when stored as recommended.
- Pakn:2 mL
- Beskrivelse:Q4ever™ Green
- Details:1250X DMSO solution
- Cat. No.:AATB17609
- Ansøgning:qPCR probe