Orders LinkContactUsLinkComponent
Q4ever™ Green
Q4ever™ Green
Q4ever™ Green, 1250X DMSO solution, 100 µl
Katalog # AATB17608
Leverandør:   AAT Bioquest
Q4ever™ Green
Katalog # AATB17608
Leverandør:  AAT Bioquest
CAS nummer:  
Q4ever™ Green
Q4ever™ Green
Q4ever™ Green, 1250X DMSO solution, 100 µl
CAS nummer:  
Price
1.620,00 kr
Log ind for tilgængelighed
1 * 100 µl
Product Details & Documents
A real-time polymerase chain reaction (real-time PCR), also known as quantitative polymerase chain reaction (qPCR), is a popular laboratory technique of molecular biology based on the polymerase chain reaction (PCR).
  • No probe required
  • Enhanced sensitivity
  • Low enzymatic inhibition
  • Binds to any dsDNA
 : For research use only. Store at −20 °C, protected from light. Product is stable for at least 12 months from the date of receipt when stored as recommended.
Specifikationer
Specifikationer
  • Pakn:
    100 µl
  • Beskrivelse:
    Q4ever™ Green
  • Details:
    1250X DMSO solution
  • Cat. No.:
    AATB17608
  • Ansøgning:
    qPCR probe
Specifikationer