Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
iFluor® 710, Succinimidyl Ester

iFluor® 710, Succinimidyl Ester

10 mg
Katalog # AATB71570
Leverandør:   AAT Bioquest
iFluor® 710, Succinimidyl Ester
Katalog # AATB71570
Leverandør:  AAT Bioquest
CAS nummer:  
iFluor® 710, Succinimidyl Ester
iFluor® 710, Succinimidyl Ester

10 mg

CAS nummer:  
Price
13.700,00 kr
Log ind for tilgængelighed
1 * 10 mg
Product Details & Documents

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal

Advarsel: For research use only.

  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakn:
    10 mg

Specifikationer