Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

iFluor® 710, Succinimidyl Ester

5 mg
Catalog # AATB71520
Supplier:   AAT Bioquest
iFluor® 710, Succinimidyl Ester
Catalog # AATB71520
Supplier:  AAT Bioquest
CAS Number:  
iFluor® 710, Succinimidyl Ester
iFluor® 710, Succinimidyl Ester

5 mg

CAS Number:  
Price
8.220,00 kr
Log in for availability
1 * 5 mg
Product Details & Documents

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal

 : For research use only.

  • Storage Temperature:Fryser
Specifications

Specifications

  • Pakn:
    5 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.