Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
iFluor® 710, Succinimidyl Ester

iFluor® 710, Succinimidyl Ester

100 µG
Katalog # AATB71045
Leverandør:   AAT Bioquest
iFluor® 710, Succinimidyl Ester
Katalog # AATB71045
Leverandør:  AAT Bioquest
CAS nummer:  
iFluor® 710, Succinimidyl Ester
iFluor® 710, Succinimidyl Ester

100 µG

CAS nummer:  
Price
872,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.

  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal

Advarsel: For research use only.

  • Opbevaringstemperatur:Fryser
Specifikationer

Specifikationer

  • Pakn:
    100 µG

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.